Learn More
Aviva Systems Biology Corp ARHGDIG Antibody - N-terminal region (ARP33854_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33854_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGDIG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ARHGDIG. Alternate names: RHOGDI-3ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-55174