Learn More
Aviva Systems Biology Corp ARHGEF1 Antibody - N-terminal region (ARP63316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGEF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human ARHGEF1. Alternate names: GEF1, LBCL2, LSC, P115-RHOGEF, SUB1.5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-937