Learn More
Aviva Systems Biology Corp ARHGEF2 Antibody - C-terminal region (ARP59669_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59669_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARHGEF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ARHGEF2. Alternate names: DKFZp547L106, DKFZp547P1516, FLJ45440, GEF, GEF-H1, GEFH1, KIAA0651, LFP40, P40ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-773