Learn More
Medchemexpress LLC Ari-3144 | 1426305-25-0 | 99.7% | 498.53 | 100 MG
SDP
Catalog No. 5000394165
Encompass_Preferred
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein (FAP).
  • Usually coupled with 7-Amino-4-methylcoumarin (AMC) for detection and quantification of FAP.
  • For research use only.
Catalog No. 50-003-94165 Supplier Medchemexpress LLC Supplier No. HY160289100MG
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein (FAP).
  • Usually coupled with 7-Amino-4-methylcoumarin (AMC) for detection and quantification of FAP.
  • For research use only.