Learn More
Medchemexpress LLC ARI-3144 | 1426305-25-0 | 99.7% | 498.53 | 25 MG
SDP
Catalog No. 5000394609
Encompass_Preferred
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (HY-D0027) (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein (FAP).
  • Can be coupled with 7-amino-4-methylcoumarin for detection and quantification of FAP.
Catalog No. 50-003-94609 Supplier Medchemexpress LLC Supplier No. 25MGHY160289
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (HY-D0027) (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein (FAP).
  • Can be coupled with 7-amino-4-methylcoumarin for detection and quantification of FAP.