Learn More
Medchemexpress LLC ARI-3144 | 1426305-25-0 | 99.7% | 498.53 | 50 MG
SDP
Catalog No. 5000393972
Encompass_Preferred
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein (FAP).
  • Usually coupled with 7-Amino-4-methylcoumarin (AMC) for FAP detection and quantification.
  • For research use only.
Catalog No. 50-003-93972 Supplier Medchemexpress LLC Supplier No. HY16028950MG
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein (FAP).
  • Usually coupled with 7-Amino-4-methylcoumarin (AMC) for FAP detection and quantification.
  • For research use only.