Learn More
Medchemexpress LLC Ari-3144 | 1426305-25-0 | 99.7% | 498.53 g/mol | C28H26N4O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16028910MG
Encompass_Preferred
ARI-3144 is a fluorogenic substrate for fibroblast activation protein (FAP) used to detect and quantify FAP enzymatic activity in biochemical assays. The compound is provided as a high-purity solid and is typically coupled to 7-amino-4-methylcoumarin (AMC) to generate a fluorescent signal upon enzymatic cleavage.
  • Fluorogenic substrate for FAP assays.
  • Compatible with AMC-based fluorescence detection.
  • High purity (>99.7%) suitable for analytical applications.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Powder stable at -20°C for long-term storage.
Catalog No. 50-002-85559