Learn More
Medchemexpress LLC ARI-3144 | 1426305-25-0 | 99.72% | 498.53 | 5 MG
SDP
Catalog No. 5000394372
Encompass_Preferred
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein.
  • Used for detection and quantification of FAP.
Catalog No. 50-003-94372 Supplier Medchemexpress LLC Supplier No. HY1602895MG
ARI-3144 is an excellent and specific substrate for fibroblast activation protein (FAP). It is typically coupled with the fluorophore 7-Amino-4-methylcoumarin (AMC) for the detection and quantification of FAP.
  • Excellent and specific substrate for fibroblast activation protein.
  • Used for detection and quantification of FAP.