Learn More
BD Cell Analysis 3P BD Biosciences Pinch Valve Tubing Assembly, For BD FACSAria Instruments, 6 mm Tubing Diameter, 1 EA
Catalog No. NC0968004
Encompass
The BD Biosciences Pinch Valve Tubing Assembly is essential for precise fluid control in BD FACSAria cell sorters, ensuring reliable sorting performance. Designed for research use only, it integrates smoothly with the fluidics system to enhance functionality compared to alternative products. This assembly is crucial for maintaining optimal conditions during cell analysis and sorting processes.
Catalog No. NC0968004 Supplier BD Cell Analysis 3P Supplier No. 65042607
The BD Biosciences Pinch Valve Tubing Assembly is essential for precise fluid control in BD FACSAria cell sorters, ensuring reliable sorting performance. Designed for research use only, it integrates smoothly with the fluidics system to enhance functionality compared to alternative products. This assembly is crucial for maintaining optimal conditions during cell analysis and sorting processes.