Learn More
BD Cell Analysis 3P BD Biosciences Waste Tank Sensor, 6-Level Monitoring, Alarm Signal Generation
Supplier:  BD Cell Analysis 3P 334915
Encompass
The BD Biosciences Waste Tank Sensor is a 6-level monitoring device designed for BD FACSAria II and FACSCanto series flow cytometers. It alerts users when waste fluid reaches critical levels, preventing overflow and protecting the instruments from damage. With a 10.6 in stem length and 24V operating voltage, it is easy to install and maintain, ensuring reliable operation in research settings. This sensor is essential for safe laboratory practices.
Catalog No. NC0620315