Learn More
Aviva Systems Biology Corp ARID3A Antibody - N-terminal region (ARP38587_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38587_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ARID3A. Alternate names: BRIGHT, DRIL1, DRIL3, E2FBP1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-56715