Learn More
Aviva Systems Biology Corp ARID3A Antibody - middle region (ARP32869_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32869_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARID3A. Alternate names: DRIL1, DRIL3, BRIGHT, E2FBP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54801