Learn More
Aviva Systems Biology Corp ARID3B Antibody - N-terminal region (ARP34275_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34275_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ARID3B. Alternate names: BDP, DRIL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52947