Learn More
Aviva Systems Biology Corp ARID3B Antibody - C-terminal region (ARP38798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ARID3B. Alternate names: BDP, DRIL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56804