Learn More
Aviva Systems Biology Corp ARID4A Antibody - C-terminal region (ARP33088_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33088_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ARID4A. Alternate names: RBP1, RBBP1, RBP-1, RBBP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54878