Learn More
Aviva Systems Biology Corp ARID5A Antibody - C-terminal region (ARP50912_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50912_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ARID5A. Alternate names: MRF-1, RP11-363D14, MRF1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60466