Learn More
Aviva Systems Biology Corp ARID5B Antibody - C-terminal region (ARP50705_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50705_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARID5B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ARID5B. Alternate names: DESRT, FLJ21150, MRF2, MRF-2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-529