Learn More
MSC Wiley X Aries Flight Gloves, X-Large, Coyote Brown, 1 Pair
Catalog No. 5000684319
Encompass
Aries flight gloves offer lightweight flame and cut protection with a Nomex/Kevlar weave on the fingers and back of hand. They feature flame-resistant treated sheepskin for enhanced dexterity and durability, along with leather reinforcement in high-wear areas and an adjustable cuff for a custom fit.
  • Lightweight Nomex/Kevlar weave for flame and cut resistance
  • Flame resistant treated sheepskin for increased dexterity and durability
  • Leather reinforced in high-wear areas
  • Adjustable cuff for custom comfort
  • NAVAIR approved and meets military flame resistance requirements
  • Provides thermal and cut resistant protection
  • Complies with ASTM D 6413, ASTM F 1790, and NFPA 1971-07 standards
  • Includes a clip strap for easy storage and portability
Catalog No. 50-006-84319 Supplier MSC Supplier No. 75147850
Aries flight gloves offer lightweight flame and cut protection with a Nomex/Kevlar weave on the fingers and back of hand. They feature flame-resistant treated sheepskin for enhanced dexterity and durability, along with leather reinforcement in high-wear areas and an adjustable cuff for a custom fit.
  • Lightweight Nomex/Kevlar weave for flame and cut resistance
  • Flame resistant treated sheepskin for increased dexterity and durability
  • Leather reinforced in high-wear areas
  • Adjustable cuff for custom comfort
  • NAVAIR approved and meets military flame resistance requirements
  • Provides thermal and cut resistant protection
  • Complies with ASTM D 6413, ASTM F 1790, and NFPA 1971-07 standards
  • Includes a clip strap for easy storage and portability