Learn More
Aviva Systems Biology Corp ARIH2 Antibody - N-terminal region (ARP34418_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34418_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARIH2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ARIH2. Alternate names: ARI2, FLJ10938, FLJ33921, TRIAD1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55384