Learn More
Aviva Systems Biology Corp ARIH2 Antibody - N-terminal region (ARP57935_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57935_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARIH2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ARIH2. Alternate names: ARI2, FLJ10938, FLJ33921, TRIAD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-415