Learn More
Medchemexpress LLC Arimoclomol citrate | 368860-21-3 | 99.8% | 505.90 g·mol⁻¹ | C20H28ClN3O10 | 1 ML
SDP
Catalog No. 5000240490
Encompass_Preferred
Arimoclomol citrate is the citrate salt of arimoclomol, a heat-shock protein (HSP) co-inducer used in research to enhance Hsp expression, reduce protein aggregation, and study neurodegenerative conditions such as Niemann-Pick disease type C. Supplied for in vitro and preclinical laboratory work, it is available as a solution in DMSO and in multiple solid pack sizes.
  • High reported purity (99.8%).
  • Supplied as a 10 mM solution in DMSO for ready use.
  • Also available in solid form for flexible dosing and formulation.
  • Molecular weight 505.90 g·mol⁻¹ for stoichiometric calculations.
  • Facilitates studies of Hsp induction and protein aggregation.
  • Suitable for in vitro and preclinical model applications.
Catalog No. 50-002-40490 Supplier Medchemexpress LLC Supplier No. HY106443B10MM 1ML
Arimoclomol citrate is the citrate salt of arimoclomol, a heat-shock protein (HSP) co-inducer used in research to enhance Hsp expression, reduce protein aggregation, and study neurodegenerative conditions such as Niemann-Pick disease type C. Supplied for in vitro and preclinical laboratory work, it is available as a solution in DMSO and in multiple solid pack sizes.
  • High reported purity (99.8%).
  • Supplied as a 10 mM solution in DMSO for ready use.
  • Also available in solid form for flexible dosing and formulation.
  • Molecular weight 505.90 g·mol⁻¹ for stoichiometric calculations.
  • Facilitates studies of Hsp induction and protein aggregation.
  • Suitable for in vitro and preclinical model applications.