Learn More
Medchemexpress LLC Arimoclomol citrate 5mg | 368860-21-3 | 5MG
SDP
Catalog No. 5000240987
Encompass_Preferred
Arimoclomol citrate (BRX-220 citrate) is an orally active co-inducer of heat shock proteins (HSP)[1 Arimoclomol citrate protects motor neurons by enhancing Hsp expression thus directly affecting protein aggregation and clearance of misfolded assemblies via the proteasome-ubiquitin system[2 Arimoclomol citrate can be used for the study of Niemann Pick disease type C
Catalog No. 50-002-40987 Supplier Medchemexpress LLC Supplier No. HY106443B5MG
Arimoclomol citrate (BRX-220 citrate) is an orally active co-inducer of heat shock proteins (HSP)[1 Arimoclomol citrate protects motor neurons by enhancing Hsp expression thus directly affecting protein aggregation and clearance of misfolded assemblies via the proteasome-ubiquitin system[2 Arimoclomol citrate can be used for the study of Niemann Pick disease type C