Learn More
Medchemexpress LLC HY-106443A 10mg Medchemexpress, Arimoclomol maleate CAS:289893-26-1 Purity:>98%
SDP
Catalog No. 501968344
Encompass_Preferred
Medchemexpress, HY-106443A 10mg Arimoclomol maleate CAS:289893-26-1 Arimoclomol maleate (BRX-220) is a co-inducer of heat shock proteins (HSP). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8344 Supplier Medchemexpress LLC Supplier No. HY106443A10MG
Medchemexpress, HY-106443A 10mg Arimoclomol maleate CAS:289893-26-1 Arimoclomol maleate (BRX-220) is a co-inducer of heat shock proteins (HSP). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.