Learn More
Medchemexpress LLC Arimoclomol maleate | 289893-26-1 | MFCD28143527 | 100.0% | 429.85 | C18H24ClN3O7 | 100 MG
SDP
Catalog No. 5000240705
Encompass_Preferred
Arimoclomol maleate is an orally active co-inducer of heat shock proteins (HSP) used in research to enhance Hsp expression and investigate proteostasis and neuroprotective mechanisms in cell and animal models. It is supplied as a high-purity solid suitable for biochemical and pharmacological studies.
  • Orally active co-inducer of heat shock proteins
  • High purity (99.96%)
  • Available in common research pack sizes including 100 mg
  • Solid, white to light yellow appearance
  • Stable under recommended storage conditions
  • Used in neuroscience and neurodegeneration research
Catalog No. 50-002-40705 Supplier Medchemexpress LLC Supplier No. HY106443A100MG
Arimoclomol maleate is an orally active co-inducer of heat shock proteins (HSP) used in research to enhance Hsp expression and investigate proteostasis and neuroprotective mechanisms in cell and animal models. It is supplied as a high-purity solid suitable for biochemical and pharmacological studies.
  • Orally active co-inducer of heat shock proteins
  • High purity (99.96%)
  • Available in common research pack sizes including 100 mg
  • Solid, white to light yellow appearance
  • Stable under recommended storage conditions
  • Used in neuroscience and neurodegeneration research