Learn More
Medchemexpress LLC Arimoclomol maleate | 289893-26-1 | MFCD28143527 | 100.0% | 429.85 | C18H24ClN3O7 | 50 MG
SDP
Catalog No. 5000240680
Encompass_Preferred
Arimoclomol maleate is the maleate salt of arimoclomol, a small-molecule co-inducer of heat shock proteins used in research on proteostasis and neurodegenerative disease models. The compound is supplied as a solid high-purity material with recommended storage and handling information provided in its datasheet and safety data sheet.
  • High purity suitable for research applications.
  • Orally active co-inducer of heat shock proteins.
  • Supplied as a stable solid with defined storage conditions.
  • Applicable to studies of neurodegeneration and proteostasis.
  • Datasheet and safety data sheet provided for handling guidance.
Catalog No. 50-002-40680 Supplier Medchemexpress LLC Supplier No. HY106443A50MG
Arimoclomol maleate is the maleate salt of arimoclomol, a small-molecule co-inducer of heat shock proteins used in research on proteostasis and neurodegenerative disease models. The compound is supplied as a solid high-purity material with recommended storage and handling information provided in its datasheet and safety data sheet.
  • High purity suitable for research applications.
  • Orally active co-inducer of heat shock proteins.
  • Supplied as a stable solid with defined storage conditions.
  • Applicable to studies of neurodegeneration and proteostasis.
  • Datasheet and safety data sheet provided for handling guidance.