Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arimoclomol maleate | 289893-26-1 | MFCD28143527 | ≥98.0% | 429.85 | C18H24ClN3O7 | 1 ML
SDP

Catalog No. 5000240491
Encompass_Preferred

Arimoclomol maleate is the maleate salt of arimoclomol (BRX-220), an orally active co-inducer of heat shock proteins used in neuroscience and neurodegeneration research. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research; solid quantities are also available for custom preparation.

  • Co-induces heat shock proteins to enhance protein quality control.
  • Provided as a 10 mM solution in DMSO, 1 mL ready stock.
  • Maleate salt form for improved stability and handling.
  • High purity (≥98.0%).
  • Molecular weight 429.85 g/mol and CAS 289893-26-1 for unambiguous identification.
  • Storage: sealed at 4°C; in solution store at -80°C up to 6 months, -20°C up to 1 month.
  • Available in solid masses (1 mg, 5 mg, 10 mg) for custom concentration preparation.

Catalog No. 50-002-40491 Supplier Medchemexpress LLC Supplier No. HY106443A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arimoclomol maleate is the maleate salt of arimoclomol (BRX-220), an orally active co-inducer of heat shock proteins used in neuroscience and neurodegeneration research. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for in vitro research; solid quantities are also available for custom preparation.

  • Co-induces heat shock proteins to enhance protein quality control.
  • Provided as a 10 mM solution in DMSO, 1 mL ready stock.
  • Maleate salt form for improved stability and handling.
  • High purity (≥98.0%).
  • Molecular weight 429.85 g/mol and CAS 289893-26-1 for unambiguous identification.
  • Storage: sealed at 4°C; in solution store at -80°C up to 6 months, -20°C up to 1 month.
  • Available in solid masses (1 mg, 5 mg, 10 mg) for custom concentration preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.