Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ ARIO Glycine Reagent, 10%

Catalog No. 01549789 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-789 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-789 Supplier Lovibond™ Supplier No. 532210
Add to Cart
Add to Cart
This item is not returnable. View return policy

The VARIO Glycine Reagent is used to measure Chlorine Dioxide in combination with Lovibond™ DPD Powder Pack reagents.

  • The VARIO product line was formulated for use in Lovibond™ test kits and instruments but can also be used in instruments and test kits from other manufacturers.

Specifications

Analytes Chlorine Dioxide
For Use With (Application) Detecting Chlorine Dioxide in a Water Sample
For Use With (Equipment) Select Lovibond™ Colorimeters and Spectrophotometers
No. of Tests ∼100
No. per Pack 29 mL
Quantity 1 Ea.
Range 0.04 to 3.8 mg/L ClO2
Sample Size 10 mL
Sterility Non-sterile
Test Method Lovibond™ M122
Test Time <5 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.