Learn More
Cayman Chemical ArIpIprazole 10mg
SDP
Catalog No. 502983428
Encompass_Preferred
An atypical antipsychotic; a partial agonist at dopamine D2, D2L, and D3 receptors (Kis = 3.3, 0.74, 9.7 nM, respectively), a partial agonist of 5-HT1A receptors (Ki = 5.6 nM), and an inverse agonist of 5-HT2B receptors (Ki = 0.36 nM); inhibits disruptions in prepulse inhibition induced by phencyclidine in mice when administered at doses of 5 and 10 mg/kg
Catalog No. 50-298-3428 Supplier Cayman Chemical Supplier No. 1998910 MG
An atypical antipsychotic; a partial agonist at dopamine D2, D2L, and D3 receptors (Kis = 3.3, 0.74, 9.7 nM, respectively), a partial agonist of 5-HT1A receptors (Ki = 5.6 nM), and an inverse agonist of 5-HT2B receptors (Ki = 0.36 nM); inhibits disruptions in prepulse inhibition induced by phencyclidine in mice when administered at doses of 5 and 10 mg/kg