Learn More
Medchemexpress LLC Aripiprazole | 129722-12-9 | 99.95% | 200 MG
SDP
Catalog No. 5000356031
Encompass_Preferred
Aripiprazole is an atypical antipsychotic that acts as a potent and high-affinity dopamine D2 receptor partial agonist. It also functions as an inverse agonist at 5-HT2B and 5-HT2A receptors and exhibits partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. This compound is suitable for research into conditions such as schizophrenia and COVID19.
  • Atypical antipsychotic
  • Potent and high-affinity dopamine D2 receptor partial agonist
  • Acts as an inverse agonist at 5-HT2B and 5-HT2A receptors
  • Exhibits partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors
  • Can be used for the research of schizophrenia and COVID19
Catalog No. 50-003-56031 Supplier Medchemexpress LLC Supplier No. HY14546200MG
Aripiprazole is an atypical antipsychotic that acts as a potent and high-affinity dopamine D2 receptor partial agonist. It also functions as an inverse agonist at 5-HT2B and 5-HT2A receptors and exhibits partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors. This compound is suitable for research into conditions such as schizophrenia and COVID19.
  • Atypical antipsychotic
  • Potent and high-affinity dopamine D2 receptor partial agonist
  • Acts as an inverse agonist at 5-HT2B and 5-HT2A receptors
  • Exhibits partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors
  • Can be used for the research of schizophrenia and COVID19