Learn More
Medchemexpress LLC Aripiprazole-d8 | 1089115-06-9 | MFCD08460937 | 98.4% | 456.43 g/mol | C23H19D8Cl2N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14546S5MG
Encompass_Preferred
Aripiprazole-d8 is the deuterium-labeled analogue of aripiprazole supplied for research use as a stable isotope reference standard. It is provided as a high-purity, small-mass sample intended for use as an internal standard in mass spectrometry and for analytical method development and pharmacokinetic studies.
  • Stable isotope-labeled internal standard for LC-MS/MS.
  • High purity suitable for analytical applications.
  • Small 5 mg sample size for solution preparation and method development.
  • Supports pharmacokinetic and metabolite studies.
  • Suitable for quantitative and qualitative mass spectrometry assays.
Catalog No. 50-002-21644