Learn More
Apexbio Technology LLC ARIPIPRAZOLE-D8-5MG
SDP
Catalog No. 5000568936
Encompass_Preferred
Aripiprazole-d8 (CAS No 1089115-06-9) is a deuterated derivative of aripiprazole used in biomedical research primarily in pharmacokinetic and pharmacodynamic studies Originating as a tool compound it plays a role in understanding the mechanism of action related to dopamine D2 and serotonin 5-HT1A receptor modulation Common in vitro activities are noted in the nanomolar to low micromolar range reflecting its effectiveness in various binding assays It is utilized in cell models and animal studies serving as a valuable asset in drug screening to assess metabolic stability and bioavailability Experimental concentrations typically depend on the specific research design allowing flexibility in study parameters
Catalog No. 50-005-68936 Supplier Apexbio Technology LLC Supplier No. SL10565
Aripiprazole-d8 (CAS No 1089115-06-9) is a deuterated derivative of aripiprazole used in biomedical research primarily in pharmacokinetic and pharmacodynamic studies Originating as a tool compound it plays a role in understanding the mechanism of action related to dopamine D2 and serotonin 5-HT1A receptor modulation Common in vitro activities are noted in the nanomolar to low micromolar range reflecting its effectiveness in various binding assays It is utilized in cell models and animal studies serving as a valuable asset in drug screening to assess metabolic stability and bioavailability Experimental concentrations typically depend on the specific research design allowing flexibility in study parameters