Learn More
AdipoGen Aripiprazole Impurity
SDP
Supplier:  AdipoGen CDXB0697G001
Encompass_Preferred
Chemical. CAS 129722-34-5. Formula C13H16BrNO2. MW 298.18. Aripiprazole is an atypical antipsychotic. Aripiprazole is a partial agonist at dopamine D2 receptors, partial agonist at 5-HT1A receptors and an antagonist at 5-HT2A, receptors. 7-4-Bromobutoxy-3,4-dihydroquinolin-21H-one is an intermediate used for the synthesis of Aripirazole. This compound can be used as a reference compound for this impurity and potential degradation product.
Catalog No. 50-256-1572