Learn More
Medchemexpress LLC Aripiprazole lauroxil | 1259305-29-7 | 99.49% | 660.71 | 1 MG
SDP
Catalog No. 5000346353
Encompass_Preferred
Aripiprazole lauroxil is an N-acyloxymethyl proagent of Aripiprazole, functioning as a long-acting injectable (LAI) antipsychotic for schizophrenia and a ligand of dopamine receptor D2R/D4R. It is cleaved by the body's enzyme esterase into active forms, with no reported toxicity. This product is intended for research use only.
  • Cleaved by the body's enzyme esterase
  • Functions as a dopamine receptor D2R/D4R ligand
  • Suitable for research applications
  • Exhibits a clearance: 0.32 ± 0.11 L/h/kg in animal models
Catalog No. 50-003-46353 Supplier Medchemexpress LLC Supplier No. HY10875150MG
Aripiprazole lauroxil is an N-acyloxymethyl proagent of Aripiprazole, functioning as a long-acting injectable (LAI) antipsychotic for schizophrenia and a ligand of dopamine receptor D2R/D4R. It is cleaved by the body's enzyme esterase into active forms, with no reported toxicity. This product is intended for research use only.
  • Cleaved by the body's enzyme esterase
  • Functions as a dopamine receptor D2R/D4R ligand
  • Suitable for research applications
  • Exhibits a clearance: 0.32 ± 0.11 L/h/kg in animal models