Learn More
Enzo Life Sciences Aristolochic acid (25mg). CAS: 313-67-7 (Aristolochic acid I), 475-80-9 (Aristolochic acid II), 67123-64-2 (mixture)
SDP
Supplier:  Enzo Life Sciences ALX270047M025
Encompass_Preferred
Phospholipase A2 (PLA2) inhibitor active against the enzymes found in many snake venoms as well as those of human platelets and synovial fluids. Inhibits ionophore-stimulated PLA2 activity in human neutrophils. Has been shown to be nephropathic and carcinogenic. Alternative name: 8-Methoxy-6-nitrophenanthrol(3 4-d)-1 3-dioxide-5-carboxylic acid, 6-Nitrophenanthrol(3 4-d)-1 3-dioxide-5-carboxylic acid. Formula: C17H11NO7 (Aristolochic acid I) and C16H9NO6 (Aristolochic acid II). MW: 341.3 (Aristolochic acid I) and 311.3 (Aristolochic acid II). Purity: ≥97% (~1:1 mixture of aristolochic acids I (A) and II (B)). Solubility: Soluble in DMSO or 100% ethanol. Sparingly soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-8380