Learn More
TARGETMOL CHEMICALS INC Aristolochic acid A 100MG
SDP
Catalog No. 502876527
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Aristolochic acid A (TR 1736) is a compound of mutagenic, carcinogenic, and nephrotoxic compounds commonly found in the Birthwort (Aristolochiaceae) family of plants. Purity 99.97%
Catalog No. 50-287-6527 Supplier TARGETMOL CHEMICALS INC Supplier No. T2801100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Aristolochic acid A (TR 1736) is a compound of mutagenic, carcinogenic, and nephrotoxic compounds commonly found in the Birthwort (Aristolochiaceae) family of plants. Purity 99.97%