Learn More
Medchemexpress LLC HY-N0510 10mg Medchemexpress, Aristolochic acid A CAS:313-67-7 Purity:>98%
SDP
Catalog No. 501933599
Encompass_Preferred
Medchemexpress, HY-N0510 10mg Aristolochic acid A CAS:313-67-7 Aristolochic acid A (Aristolochic acid I; TR 1736) is the main component of plant extract Aristolochic acids, which are found in various herbal plants of genus Aristolochia and Asarum. Aristolochic acid A significantly reduces both activator protein 1 (AP-1) and NF-κB activities. Aristolochic acid A reduces BLCAP gene expression in human cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3599 Supplier Medchemexpress LLC Supplier No. HYN051010MG
Medchemexpress, HY-N0510 10mg Aristolochic acid A CAS:313-67-7 Aristolochic acid A (Aristolochic acid I; TR 1736) is the main component of plant extract Aristolochic acids, which are found in various herbal plants of genus Aristolochia and Asarum. Aristolochic acid A significantly reduces both activator protein 1 (AP-1) and NF-κB activities. Aristolochic acid A reduces BLCAP gene expression in human cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.