Learn More
Apexbio Technology LLC ARISTOLOCHIC-ACID-A-20MG
SDP
Catalog No. 5000567950
Encompass_Preferred
Aristolochic Acid A (CAS No 313-67-7) is a compound primarily explored in the field of toxicology and pharmacology Originating from the Aristolochiaceae family it is known for its role as a potent nephrotoxin and carcinogen making it a subject of interest in studies of renal toxicity and cancer research The compound acts by binding to DNA forming adducts that can lead to mutations particularly affecting pathways related to the cell cycle and apoptosis Its activity is typically observed in the nanomolar to low micromolar range Aristolochic Acid A is often used in vitro with various cell models such as renal epithelial cells and in vivo in animal models to understand its toxic effects and the mechanisms leading to tumorigenesis It is also employed in drug screening processes to identify potential inhibitors that may counter its detrimental effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67950 Supplier Apexbio Technology LLC Supplier No. N203220
Aristolochic Acid A (CAS No 313-67-7) is a compound primarily explored in the field of toxicology and pharmacology Originating from the Aristolochiaceae family it is known for its role as a potent nephrotoxin and carcinogen making it a subject of interest in studies of renal toxicity and cancer research The compound acts by binding to DNA forming adducts that can lead to mutations particularly affecting pathways related to the cell cycle and apoptosis Its activity is typically observed in the nanomolar to low micromolar range Aristolochic Acid A is often used in vitro with various cell models such as renal epithelial cells and in vivo in animal models to understand its toxic effects and the mechanisms leading to tumorigenesis It is also employed in drug screening processes to identify potential inhibitors that may counter its detrimental effects Experimental concentrations typically depend on the specific research design