Learn More
Medchemexpress LLC Aristolochic acid A | 313-67-7 | 341.27 | 1 MG
SDP
Catalog No. 5000389846
Encompass_Preferred
Aristolochic acid A is the primary component of Aristolochic acids, naturally occurring in plants of the genus Aristolochia and Asarum. This compound has been shown to significantly reduce both activator protein 1 (AP-1) and NF-κB activities. It also decreases BLCAP gene expression in human cell lines, making it a valuable tool in various biological studies. Additionally, Aristolochic acid A is utilized in research to induce nephrotoxicity models.
  • Reduces activator protein 1 (AP-1) activity
  • Reduces NF-κB activity
  • Reduces BLCAP gene expression in human cell lines
  • Can be used to induce nephrotoxicity models
  • High purity (99.91%)
  • Light yellow to orange solid appearance
Catalog No. 50-003-89846 Supplier Medchemexpress LLC Supplier No. HYN0510A1MG
Aristolochic acid A is the primary component of Aristolochic acids, naturally occurring in plants of the genus Aristolochia and Asarum. This compound has been shown to significantly reduce both activator protein 1 (AP-1) and NF-κB activities. It also decreases BLCAP gene expression in human cell lines, making it a valuable tool in various biological studies. Additionally, Aristolochic acid A is utilized in research to induce nephrotoxicity models.
  • Reduces activator protein 1 (AP-1) activity
  • Reduces NF-κB activity
  • Reduces BLCAP gene expression in human cell lines
  • Can be used to induce nephrotoxicity models
  • High purity (99.91%)
  • Light yellow to orange solid appearance