Learn More
Medchemexpress LLC Aristolochic acid A | 313-67-7 | 341.27 | 5 MG
SDP
Catalog No. 5000389226
Encompass_Preferred
Aristolochic acid A is the main component of plant extract Aristolochic acids, found in various herbal plants of the genus Aristolochia and Asarum. This product is an analytical standard intended for research and analytical applications. It significantly reduces both activator protein 1 (AP-1) and NF-κB activities, and reduces BLCAP gene expression in human cell lines.
  • Reduces both activator protein 1 (AP-1) and NF-κB activities.
  • Reduces BLCAP gene expression in human cell lines.
Catalog No. 50-003-89226 Supplier Medchemexpress LLC Supplier No. HYN05105MG
Aristolochic acid A is the main component of plant extract Aristolochic acids, found in various herbal plants of the genus Aristolochia and Asarum. This product is an analytical standard intended for research and analytical applications. It significantly reduces both activator protein 1 (AP-1) and NF-κB activities, and reduces BLCAP gene expression in human cell lines.
  • Reduces both activator protein 1 (AP-1) and NF-κB activities.
  • Reduces BLCAP gene expression in human cell lines.