Learn More
Medchemexpress LLC Aristolochic acid A | 313-67-7 | MFCD00004996 | 99.9% | 10 MG
SDP
Catalog No. 5000296900
Encompass_Preferred
Aristolochic acid A is a nitrophenanthrene carboxylic acid supplied as a high-purity research reagent for biochemical and cell-based studies. It is commonly used to investigate NF-κB and AP-1 signaling pathways and for mechanistic studies of nephrotoxicity and carcinogenesis.
  • High purity (99.9%).
  • Molecular formula C17H11NO7; molecular weight 341.27 g/mol.
  • Typically supplied as a 10 mg quantity suitable for research use.
  • Used in studies of NF-κB and AP-1 inhibition and mechanisms of nephrotoxicity and carcinogenicity.
  • Handle as a toxic and carcinogenic compound with appropriate safety precautions.
Catalog No. 50-002-96900 Supplier Medchemexpress LLC Supplier No. HYN0510A10MG
Aristolochic acid A is a nitrophenanthrene carboxylic acid supplied as a high-purity research reagent for biochemical and cell-based studies. It is commonly used to investigate NF-κB and AP-1 signaling pathways and for mechanistic studies of nephrotoxicity and carcinogenesis.
  • High purity (99.9%).
  • Molecular formula C17H11NO7; molecular weight 341.27 g/mol.
  • Typically supplied as a 10 mg quantity suitable for research use.
  • Used in studies of NF-κB and AP-1 inhibition and mechanisms of nephrotoxicity and carcinogenicity.
  • Handle as a toxic and carcinogenic compound with appropriate safety precautions.