Learn More
Cayman Chemical Aristolochic Acid B, Purity: ≥98%, CAS: 475-80-9, 10 mg
SDP
Catalog No. 502939342
Encompass_Preferred
An alkaloid with mutagenic activity; forms DNA adducts under anaerobic conditions in vitro at 0.4 mM; oral administration induces bladder, forestomach, and kidney DNA adduct formation in rats at 0.03 mmol/kg; reduces body weight and urine concentrations of sodium, potassium, and calcium, enlarges the liver and kidneys, and increases blood concentrations of hemoglobin and hematocrit in rats
Catalog No. 50-293-9342 Supplier Cayman Chemical Supplier No. 2850410 MG
An alkaloid with mutagenic activity; forms DNA adducts under anaerobic conditions in vitro at 0.4 mM; oral administration induces bladder, forestomach, and kidney DNA adduct formation in rats at 0.03 mmol/kg; reduces body weight and urine concentrations of sodium, potassium, and calcium, enlarges the liver and kidneys, and increases blood concentrations of hemoglobin and hematocrit in rats