Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aristolochic acid B | 475-80-9 | 311.25 | 5 MG
SDP

Catalog No. 5000388767
Encompass_Preferred

Aristolochic acid B (Aristolochic Acid II) is an orally active major component of aristolochic acid (AA). It can be isolated from plants of the genus Aristolochica. Aristolochic acid B forms DNA adducts, is mutagenic, and exhibits a greater carcinogenic risk in vivo than Aristolochic acid I.

  • Target: DNA alkylator/crosslinker
  • Research area: Cancer
  • Mechanism: Forms DNA adducts and is mutagenic
  • Carcinogenic risk: Exhibits greater carcinogenic risk in vivo than Aristolochic acid I
  • Appearance: Solid
  • Melting/freezing point: 270-272°C

Catalog No. 50-003-88767 Supplier Medchemexpress LLC Supplier No. HYN05115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aristolochic acid B (Aristolochic Acid II) is an orally active major component of aristolochic acid (AA). It can be isolated from plants of the genus Aristolochica. Aristolochic acid B forms DNA adducts, is mutagenic, and exhibits a greater carcinogenic risk in vivo than Aristolochic acid I.

  • Target: DNA alkylator/crosslinker
  • Research area: Cancer
  • Mechanism: Forms DNA adducts and is mutagenic
  • Carcinogenic risk: Exhibits greater carcinogenic risk in vivo than Aristolochic acid I
  • Appearance: Solid
  • Melting/freezing point: 270-272°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.