Learn More
TARGETMOL CHEMICALS INC Aristolochic acid B 5MG
SDP
Catalog No. 502877897
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aristolochic acid B (Aristolochic Acid II) is a natural product from the Aristolochaceae taxon and is one of the main components of aristolochic acid. Aristolochic acid is a mutagen and carcinogen, and Aristolochic acid B (Aristolochic Acid II) can show a greater carcinogenic risk in vivo than Aristolochic acid I. Purity 99.99%
Catalog No. 50-287-7897 Supplier TARGETMOL CHEMICALS INC Supplier No. T70315MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aristolochic acid B (Aristolochic Acid II) is a natural product from the Aristolochaceae taxon and is one of the main components of aristolochic acid. Aristolochic acid is a mutagen and carcinogen, and Aristolochic acid B (Aristolochic Acid II) can show a greater carcinogenic risk in vivo than Aristolochic acid I. Purity 99.99%