Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aristolochic acid B (aristolochic acid II) | 475-80-9 | 311.25 | 1 MG
SDP

Catalog No. 5000388495
Encompass_Preferred

Aristolochic acid B (Aristolochic Acid II) is an orally active major component of aristolochic acid (AA). It can be isolated from plants of the genus Aristolochica, forms DNA adducts, and is mutagenic. It exhibits a greater carcinogenic risk in vivo than Aristolochic acid I.

  • Orally active major component of aristolochic acid.
  • Isolated from plants of the Aristolochica genus.
  • Forms DNA adducts and is mutagenic.
  • Shows greater carcinogenic risk in vivo than Aristolochic acid I.
  • Purity of 99.92%.
  • Soluble in DMSO for in vitro studies.
  • Specific storage conditions to maintain stability.

Catalog No. 50-003-88495 Supplier Medchemexpress LLC Supplier No. HYN05111MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aristolochic acid B (Aristolochic Acid II) is an orally active major component of aristolochic acid (AA). It can be isolated from plants of the genus Aristolochica, forms DNA adducts, and is mutagenic. It exhibits a greater carcinogenic risk in vivo than Aristolochic acid I.

  • Orally active major component of aristolochic acid.
  • Isolated from plants of the Aristolochica genus.
  • Forms DNA adducts and is mutagenic.
  • Shows greater carcinogenic risk in vivo than Aristolochic acid I.
  • Purity of 99.92%.
  • Soluble in DMSO for in vitro studies.
  • Specific storage conditions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.