Learn More
Cayman Chemical Aristolochic Acid C, Purity: ≥95%, CAS: 4849-90-5, 10 mg
SDP
Catalog No. 502940624
Encompass_Preferred
An alkaloid; inhibits the growth of LLC-PK1, P388, HT-29, and HL-60 cancer cells (IC50s = 85, 8.8, 17.1, and 40 µM, respectively)
Catalog No. 50-294-0624 Supplier Cayman Chemical Supplier No. 2850510 MG
An alkaloid; inhibits the growth of LLC-PK1, P388, HT-29, and HL-60 cancer cells (IC50s = 85, 8.8, 17.1, and 40 µM, respectively)