Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aristolochic acid C | 4849-90-5 | 327.25 | 1 MG
SDP

Catalog No. 5000416627
Encompass_Preferred

Aristolochic acid C is a derivative of Aristolochic acid, which functions as a phospholipase A2 (PLA2) inhibitor. This inhibition disrupts cortical microtubule arrays and root growth in *Arabidopsis*.

  • Purity: 99.77%
  • Appearance: solid
  • Color: light yellow to orange
  • Solubility in vitro: DMSO : 125 mg/mL (381.97 mM; needs ultrasonic)
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protected from light
  • Storage in solvent: -80°C, 6 months; -20°C, 1 month (protected from light)
  • In vivo dissolution protocol: 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline (solubility: ≥ 2.08 mg/mL (6.36 mM); clear solution)

Catalog No. 50-004-16627 Supplier Medchemexpress LLC Supplier No. HYN14501MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aristolochic acid C is a derivative of Aristolochic acid, which functions as a phospholipase A2 (PLA2) inhibitor. This inhibition disrupts cortical microtubule arrays and root growth in *Arabidopsis*.

  • Purity: 99.77%
  • Appearance: solid
  • Color: light yellow to orange
  • Solubility in vitro: DMSO : 125 mg/mL (381.97 mM; needs ultrasonic)
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protected from light
  • Storage in solvent: -80°C, 6 months; -20°C, 1 month (protected from light)
  • In vivo dissolution protocol: 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline (solubility: ≥ 2.08 mg/mL (6.36 mM); clear solution)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.