Learn More
Medchemexpress LLC Aristolochic acid D (Aristolochic acid-IVa) | 17413-38-6 | 357.27 | 1 MG
SDP
Catalog No. 5000414524
Encompass_Preferred
Aristolochic acid D (Aristolochic acid-IVa) is an orally active PDE2 and CDK2 inhibitor isolated from *Aristolochia indica L.* It exhibits anti-inflammatory activity and can be used in the research of inflammation and tumor-related diseases.
  • Orally active PDE2 inhibitor
  • Orally active CDK2 inhibitor
  • Isolated from *Aristolochia indica L.*
  • Exhibits anti-inflammatory activity
  • Non-carcinogenic
  • Non-nephrotoxic
  • Can be used in inflammation research
  • Can be used in tumor-related diseases research
Catalog No. 50-004-14524 Supplier Medchemexpress LLC Supplier No. HYN14651MG
Aristolochic acid D (Aristolochic acid-IVa) is an orally active PDE2 and CDK2 inhibitor isolated from *Aristolochia indica L.* It exhibits anti-inflammatory activity and can be used in the research of inflammation and tumor-related diseases.
  • Orally active PDE2 inhibitor
  • Orally active CDK2 inhibitor
  • Isolated from *Aristolochia indica L.*
  • Exhibits anti-inflammatory activity
  • Non-carcinogenic
  • Non-nephrotoxic
  • Can be used in inflammation research
  • Can be used in tumor-related diseases research