Learn More
Medchemexpress LLC Aristolochic acid D (Aristolochic acid-IVa) | 17413-38-6 | 357.27 | 1 ML
SDP
Catalog No. 5000416263
Encompass_Preferred
Aristolochic acid D (Aristolochic acid-IVa) is an orally active inhibitor of PDE2 (IC50: 4.673 μM) and CDK2 (IC50: 25 μM) that is isolated from Aristolochia indica L. It demonstrates anti-inflammatory activity and is considered non-carcinogenic and non-nephrotoxic. This compound is suitable for research related to inflammation and tumor-related diseases.
  • Orally active PDE2 inhibitor (IC50: 4.673 μM).
  • CDK2 inhibitor (IC50: 25 μM).
  • Isolated from Aristolochia indica L.
  • Exhibits anti-inflammatory activity.
  • Non-carcinogenic.
  • Non-nephrotoxic.
  • Useful in research for inflammation and tumor-related diseases.
Catalog No. 50-004-16263 Supplier Medchemexpress LLC Supplier No. HYN146510MM/1ML
Aristolochic acid D (Aristolochic acid-IVa) is an orally active inhibitor of PDE2 (IC50: 4.673 μM) and CDK2 (IC50: 25 μM) that is isolated from Aristolochia indica L. It demonstrates anti-inflammatory activity and is considered non-carcinogenic and non-nephrotoxic. This compound is suitable for research related to inflammation and tumor-related diseases.
  • Orally active PDE2 inhibitor (IC50: 4.673 μM).
  • CDK2 inhibitor (IC50: 25 μM).
  • Isolated from Aristolochia indica L.
  • Exhibits anti-inflammatory activity.
  • Non-carcinogenic.
  • Non-nephrotoxic.
  • Useful in research for inflammation and tumor-related diseases.