Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aristolochic acid III | 7267-92-7 | 1 MG
SDP

Catalog No. 5000417524
Encompass_Preferred

Aristolochic acid III is a natural product extracted from the leaves of Aristolochia debilis, offering a purity of 85.0% and a molecular weight of 341.27. It appears as a white to light yellow solid.

  • Isolated from Aristolochia debilis leaves
  • Purity: 85.0%
  • Molecular formula: C17H11NO7
  • Molecular weight: 341.27
  • Solid appearance
  • Color: White to light yellow
  • Stable under recommended storage conditions

Catalog No. 50-004-17524 Supplier Medchemexpress LLC Supplier No. HYN79511MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aristolochic acid III is a natural product extracted from the leaves of Aristolochia debilis, offering a purity of 85.0% and a molecular weight of 341.27. It appears as a white to light yellow solid.

  • Isolated from Aristolochia debilis leaves
  • Purity: 85.0%
  • Molecular formula: C17H11NO7
  • Molecular weight: 341.27
  • Solid appearance
  • Color: White to light yellow
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.