Learn More
Bollé Safety™ Arizona D28 Bifocals with Rx Lens
Catalog No. 18072141
Change view
Click to view available options
Lens Tint:
Brown
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
18-072-141 Gray
18-072-140 Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-072-141 Supplier Bollé Safety™ Supplier No. CRBPHOTOG1
This item is not returnable. View return policy
Prescription lens option for safety glasses.
  • Lenses made of CR39 plastic
  • Available in multiple colors
Specifications
Lens Coating Uncoated
Lens Type Bifocal Photochromatic
Lens Tint Gray
Product Type Prescription Lens
Disposable Reusable
Lens Material Plastic
Autoclavable Not Autoclavable